Assay Details
| Gene Symbol: LOC105372626 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134577.1 | 1 - 2 | 87 | 95 | ||
| GenBank mRNA | BX089899.1 | - | 1 - 2 | 88 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927200 | LOC101927200 | -73.8kb | Sense | Chr. 20 - 43727388 - 43820895 on NCBI Build GRCh38 | |
| Downstream | TOX high mobility group box family member 2 | TOX2 |
C20orf100, GCX-1, GCX1, dJ1108D11.2, dJ495O3.1 |
19.4kb | Sense | Chr. 20 - 43914852 - 44069616 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105372626
Genomic Map