Assay Details
| Gene Symbol: LOC105372069 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134585.1 | 2 - 3 | 190 | 102 | ||
| GenBank mRNA | BC037214.1 | - | 2 - 3 | 190 | 102 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | KIAA1328 | KIAA1328 | Anti-sense | Chr. 18 - 36828929 - 37232127 on NCBI Build GRCh38 | ||
| Downstream |
CUGBP, Elav-like family member 4 |
CELF4 |
BRUNOL4, CELF-4 |
321.8kb | Sense | Chr. 18 - 37243040 - 37566052 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105372069
Genomic Map