Assay Details
| Gene Symbol: LOC105377962 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134602.1 | 2 - 3 | 74 | 110 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | heparan sulfate-glucosamine 3-sulfotransferase 5 | HS3ST5 |
3-OST-5, 3OST5, HS3OST5, NBLA04021 |
-1.3mb | Sense | Chr. 6 - 114055586 - 114342388 on NCBI Build GRCh38 |
| Downstream | fyn related Src family tyrosine kinase | FRK |
GTK, PTK5, RAK |
304.8kb | Sense | Chr. 6 - 115941031 - 116100739 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105377962
Genomic Map