Assay Details
| Gene Symbol: LOC105374988 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134611.1 | 1 - 2 | 210 | 129 | ||
| GenBank mRNA | BC033330.1 | - | 1 - 2 | 210 | 129 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | high mobility group nucleosomal binding domain 4 | HMGN4 |
HMG17L3, NHC |
-22.4kb | Sense | Chr. 6 - 26538344 - 26546937 on NCBI Build GRCh38 |
| Downstream | activator of basal transcription 1 | ABT1 | hABT1 | 22.2kb | Sense | Chr. 6 - 26596943 - 26600050 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105374988
Genomic Map