Assay Details
| Gene Symbol: LOC105373021 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134623.1 | 2 - 3 | 237 | 110 | ||
| GenBank mRNA | DA099798.1 | - | 2 - 3 | 237 | 110 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | intraflagellar transport 27 | IFT27 |
BBS19, RABL4, RAYL |
Anti-sense | Chr. 22 - 36758202 - 36776133 on NCBI Build GRCh38 | |
| Downstream | parvalbumin | PVALB | D22S749 | 34.2kb | Anti-sense | Chr. 22 - 36800701 - 36819473 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105373021
Genomic Map