Assay Details
| Gene Symbol: LOC105373031 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134627.1 | 1 - 2 | 90 | 75 | ||
| GenBank mRNA | HY041837.1 | - | 1 - 2 | 90 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DNA meiotic recombinase 1 | DMC1 |
DMC1H, LIM15, dJ199H16.1 |
-214.0 | Anti-sense | Chr. 22 - 38509154 - 38570286 on NCBI Build GRCh38 |
| Downstream | family with sequence similarity 227 member A | FAM227A | 6.8kb | Anti-sense | Chr. 22 - 38578120 - 38656629 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105373031
Genomic Map