Assay Details
| Gene Symbol: LOC101929161 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134641.1 | 3 - 4 | 425 | 137 | ||
| GenBank mRNA | AA885339.1 | - | 1 - 2 | 137 | ||
| BX099416.1 | - | 3 - 4 | 425 | 137 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | anaphase promoting complex subunit 4 | ANAPC4 | APC4 | -112.5kb | Anti-sense | Chr. 4 - 25377226 - 25418498 on NCBI Build GRCh38 |
| Downstream | solute carrier family 34 member 2 | SLC34A2 |
NAPI-3B, NAPI-IIb, NPTIIb |
120.5kb | Anti-sense | Chr. 4 - 25655813 - 25678748 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101929161
Genomic Map