Assay Details
| Gene Symbol: LOC105372397 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134907.1 | 2 - 3 | 228 | 77 | ||
| GenBank mRNA | BM716908.1 | - | 2 - 3 | 229 | 77 | |
| BU738492.1 | - | 1 - 2 | 77 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitogen-activated protein kinase kinase kinase kinase 1 | MAP4K1 | HPK1 | Anti-sense | Chr. 19 - 38587640 - 38618035 on NCBI Build GRCh38 | |
| Downstream | eukaryotic translation initiation factor 3 subunit K | EIF3K |
ARG134, EIF3-p28, EIF3S12, HSPC029, M9, MSTP001, PLAC-24, PLAC24, PRO1474, PTD001 |
17.4kb | Sense | Chr. 19 - 38619072 - 38636959 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105372397
Genomic Map