Assay Details
| Gene Symbol: LOC101929697 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134940.1 | 1 - 2 | 98 | 88 | ||
| GenBank mRNA | DA288418.1 | - | 1 - 2 | 101 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | proline-rich protein 36-like | LOC105374299 | -17.7kb | Anti-sense | Chr. 3 - 195815706 - 195818776 on NCBI Build GRCh38 | |
| Downstream | tyrosine kinase non receptor 2 | TNK2 |
ACK, ACK-1, ACK1, p21cdc42Hs |
9.1kb | Sense | Chr. 3 - 195863364 - 195909009 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101929697
Genomic Map