Assay Details
| Gene Symbol: LOC102724231 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134943.1 | 1 - 2 | 62 | 102 | ||
| GenBank mRNA | BX377745.2 | - | 1 - 2 | 44 | 102 | |
| HY025704.1 | - | 1 - 2 | 63 | 102 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
T-cell activation inhibitor, mitochondrial |
TCAIM |
C3orf23, TOAG-1, TOAG1 |
-11.7kb | Anti-sense | Chr. 3 - 44338119 - 44409448 on NCBI Build GRCh38 |
| Downstream | zinc finger protein 445 | ZNF445 |
ZKSCAN15, ZNF168, ZSCAN47 |
15.7kb | Sense | Chr. 3 - 44439770 - 44477670 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC102724231
Genomic Map