Assay Details
| Gene Symbol: LOC105370068 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134995.1 | 2 - 3 | 138 | 93 | ||
| GenBank mRNA | AI458874.1 | - | 2 - 3 | 93 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927637 | LOC101927637 | -12.8kb | Sense | Chr. 12 - 127631269 - 127636467 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC400087 | FLJ37505 | 230.3kb | Anti-sense | Chr. 12 - 127881617 - 127898639 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105370068
Genomic Map