Assay Details
| Gene Symbol: LOC105370016 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_134997.1 | 1 - 2 | 147 | 71 | ||
| GenBank mRNA | AL037786.1 | - | 1 - 2 | 151 | 71 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC105370014 | LOC105370014 | -71.3kb | Sense | Chr. 12 - 118758754 - 118761679 on NCBI Build GRCh38 | |
| Downstream | serine/arginine repetitive matrix 4 | SRRM4 |
KIAA1853, MU-MB-2.76, nSR100 |
148.0kb | Anti-sense | Chr. 12 - 118981495 - 119163051 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105370016
Genomic Map