Assay Details
| Gene Symbol: PTGER4P2-CDK2AP2P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135010.1 | 1 - 2 | 157 | 81 | ||
| GenBank mRNA | BC068605.1 | - | 1 - 2 | 156 | 81 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BI600443.1 | - | 161 | Chr. - - [-] on Build | 81 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC100132249 | LOC100132249 | -2.3kb | Anti-sense | Chr. 9 - 62834248 - 62838346 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC403323 | LOC403323 | 13.7kb | Anti-sense | Chr. 9 - 62857856 - 62898087 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900010233.hg.1 | ENST00000458150.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007364.hg.1 | ENST00000328411.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007364.hg.1 | ENST00000478512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007364.hg.1 | NR_024496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007364.hg.1 | NR_024496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007364.hg.1 | NR_024496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007364.hg.1 | NR_024496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007364.hg.1 | NR_024496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007364.hg.1 | NR_024496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007364.hg.1 | NR_024496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007364.hg.1 | NR_024496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007364.hg.1 | NR_024496.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007265.hg.1 | ENST00000617479.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007265.hg.1 | ENST00000619442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007265.hg.1 | ENST00000619442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007265.hg.1 | ENST00000619442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007265.hg.1 | ENST00000619442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007265.hg.1 | ENST00000619442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007265.hg.1 | ENST00000619442.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900010232.hg.1 | ENST00000422476.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000609782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000609782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000609782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000609782.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000631186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000631186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000631186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | ENST00000631186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007265.hg.1 | ENST00000617479.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007265.hg.1 | ENST00000619442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007265.hg.1 | ENST00000619442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007265.hg.1 | ENST00000619442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007265.hg.1 | ENST00000619442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007265.hg.1 | ENST00000619442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007265.hg.1 | ENST00000619442.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007364.hg.1 | ENST00000328411.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007364.hg.1 | ENST00000478512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007364.hg.1 | NR_024496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007364.hg.1 | NR_024496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007364.hg.1 | NR_024496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007364.hg.1 | NR_024496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007364.hg.1 | NR_024496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007364.hg.1 | NR_024496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007364.hg.1 | NR_024496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007364.hg.1 | NR_024496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007364.hg.1 | NR_024496.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000423808.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000426534.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000436878.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000458316.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000609782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000609782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000609782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000609782.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000616647.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000631186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000631186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000631186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | ENST00000631186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126010.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100006834.hg.1 | NR_126011.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900010232.hg.1 | ENST00000422476.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900010233.hg.1 | ENST00000458150.2 | Ensembl |

PTGER4P2-CDK2AP2P2
Genomic Map