Assay Details
| Gene Symbol: LOC105369891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135019.1 | 1 - 2 | 54 | 96 | ||
| GenBank mRNA | BX090745.1 | - | 1 - 2 | 54 | 96 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 936 | LINC00936 | -235.7kb | Sense | Chr. 12 - 89708955 - 89711952 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC105369893 | LOC105369893 | 345.1kb | Sense | Chr. 12 - 90293318 - 90300698 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105369891
Genomic Map