Assay Details
| Gene Symbol: LOC105369860 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135021.1 | 1 - 2 | 565 | 98 | ||
| GenBank mRNA | BC034620.1 | - | 1 - 2 | 537 | 98 | |
| BC045810.1 | - | 1 - 2 | 565 | 98 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neuron navigator 3 | NAV3 |
POMFIL1, STEERIN3, unc53H3 |
-113.7kb | Anti-sense | Chr. 12 - 77571862 - 78213012 on NCBI Build GRCh38 |
| Downstream | synaptotagmin 1 | SYT1 |
P65, SVP65, SYT |
504.2kb | Anti-sense | Chr. 12 - 78863993 - 79452008 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105369860
Genomic Map