Assay Details
| Gene Symbol: LOC105369781 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135023.1 | 2 - 3 | 205 | 80 | ||
| GenBank mRNA | HY183314.1 | - | 1 - 2 | 80 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | IKAROS family zinc finger 4 | IKZF4 |
EOS, ZNFN1A4 |
Anti-sense | Chr. 12 - 56007647 - 56038435 on NCBI Build GRCh38 | |
| Downstream | ribosomal protein S26 | RPS26 |
DBA10, S26 |
30.2kb | Anti-sense | Chr. 12 - 56041902 - 56044223 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105369781
Genomic Map