| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007395.hg.1 |
ENST00000546982.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007395.hg.1 |
ENST00000546982.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007395.hg.1 |
ENST00000546982.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007395.hg.1 |
ENST00000546982.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007395.hg.1 |
ENST00000546982.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007395.hg.1 |
ENST00000548764.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007395.hg.1 |
ENST00000548764.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007395.hg.1 |
ENST00000548764.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007395.hg.1 |
ENST00000548764.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007395.hg.1 |
ENST00000548764.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007395.hg.1 |
ENST00000548764.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007395.hg.1 |
ENST00000548764.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007395.hg.1 |
ENST00000548764.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007395.hg.1 |
ENST00000548764.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1200007395.hg.1 |
ENST00000548764.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007395.hg.1 |
ENST00000546982.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007395.hg.1 |
ENST00000546982.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007395.hg.1 |
ENST00000546982.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007395.hg.1 |
ENST00000546982.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007395.hg.1 |
ENST00000546982.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007395.hg.1 |
ENST00000548764.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007395.hg.1 |
ENST00000548764.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007395.hg.1 |
ENST00000548764.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007395.hg.1 |
ENST00000548764.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007395.hg.1 |
ENST00000548764.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007395.hg.1 |
ENST00000548764.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007395.hg.1 |
ENST00000548764.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007395.hg.1 |
ENST00000548764.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007395.hg.1 |
ENST00000548764.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1200007395.hg.1 |
ENST00000548764.1 |
Ensembl |