Assay Details
| Gene Symbol: LOC105369723 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135029.1 | 1 - 2 | 40 | 64 | ||
| GenBank mRNA | BI771000.1 | - | 1 - 2 | 40 | 64 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
H3 histone, family 3C |
H3F3C | H3.5 | -84.7kb | Sense | Chr. 12 - 31791185 - 31792241 on NCBI Build GRCh38 |
| Downstream | KIAA1551 | KIAA1551 |
C12orf35, UTA2-1 |
71.6kb | Anti-sense | Chr. 12 - 31958800 - 31993107 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105369723
Genomic Map