Assay Details
| Gene Symbol: LOC101929538 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135072.1 | 1 - 2 | 693 | 68 | ||
| GenBank mRNA | AK093020.1 | - | 1 - 2 | 693 | 68 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
Fli-1 proto-oncogene, ETS transcription factor |
FLI1 |
EWSR2, SIC-1 |
Anti-sense | Chr. 11 - 128685263 - 128813267 on NCBI Build GRCh38 | |
| Downstream | smooth muscle and endothelial cell enriched migration/differentiation-associated long non-coding RNA | SENCR |
FLI1-AS1, lncRNA9 |
4.8kb | Sense | Chr. 11 - 128691672 - 128696023 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101929538
Genomic Map