Assay Details
| Gene Symbol: LOC105372897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135085.1 | 1 - 2 | 130 | 79 | ||
| GenBank mRNA | AW270068.1 | - | 2 - 3 | 79 | ||
| BX119711.1 | - | 1 - 2 | 130 | 79 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | plexin A2 | PLXNA2 |
OCT, PLXN2 |
-1.1mb | Anti-sense | Chr. 1 - 208022242 - 208244384 on NCBI Build GRCh38 |
| Downstream | MIR205 host gene | MIR205HG | LINC00510 | 101.3kb | Sense | Chr. 1 - 209428820 - 209432552 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105372897
Genomic Map