Assay Details
| Gene Symbol: LOC105371433 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135098.1 | 1 - 2 | 76 | 105 | ||
| GenBank mRNA | DA671305.1 | - | 1 - 2 | 73 | 105 | |
| DB153018.1 | - | 1 - 2 | 76 | 105 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | pleckstrin homology domain containing O1 | PLEKHO1 |
CKIP-1, CKIP1, JBP, OC120 |
-663.0 | Sense | Chr. 1 - 150149434 - 150160065 on NCBI Build GRCh38 |
| Downstream | acidic nuclear phosphoprotein 32 family member E | ANP32E |
LANP-L, LANPL |
54.1kb | Anti-sense | Chr. 1 - 150218417 - 150236156 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105371433
Genomic Map