Assay Details
| Gene Symbol: LOC105370586 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135159.1 | 1 - 2 | 61 | 94 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | aarF domain containing kinase 1 | ADCK1 | -1.3mb | Sense | Chr. 14 - 77800058 - 77933954 on NCBI Build GRCh38 | |
| Downstream |
deiodinase, iodothyronine, type II |
DIO2 |
5DII, D2, DIOII, SelY, TXDI2 |
995.9kb | Anti-sense | Chr. 14 - 80197525 - 80231054 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105370586
Genomic Map