Assay Details
| Gene Symbol: LOC105371335 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135172.1 | 1 - 2 | 120 | 109 | ||
| GenBank mRNA | HY013069.1 | - | 1 - 2 | 121 | 109 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 19 | ZNF19 | KOX12 | -24.5kb | Anti-sense | Chr. 16 - 71474073 - 71489351 on NCBI Build GRCh38 |
| Downstream | carbohydrate sulfotransferase 4 | CHST4 |
GST3, GlcNAc6ST2, HECGLCNAC6ST, LSST |
8.3kb | Sense | Chr. 16 - 71526120 - 71538590 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105371335
Genomic Map