Assay Details
| Gene Symbol: LOC101928659 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135180.1 | 1 - 2 | 218 | 55 | ||
| GenBank mRNA | BQ640484.1 | - | 1 - 2 | 215 | 55 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 16 open reading frame 95 | C16orf95 | -89.0 | Anti-sense | Chr. 16 - 87302798 - 87317420 on NCBI Build GRCh38 | |
| Downstream | F-box protein 31 | FBXO31 |
FBX14, FBXO14, Fbx31, MRT45, pp2386 |
8.9kb | Anti-sense | Chr. 16 - 87326987 - 87392107 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101928659
Genomic Map