| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007027.hg.1 |
ENST00000555433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007027.hg.1 |
ENST00000555433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007027.hg.1 |
ENST00000555433.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007027.hg.1 |
ENST00000556228.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007027.hg.1 |
ENST00000556228.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007027.hg.1 |
ENST00000557465.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007027.hg.1 |
ENST00000557465.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007027.hg.1 |
ENST00000557465.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1400007027.hg.1 |
ENST00000557465.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007027.hg.1 |
ENST00000555433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007027.hg.1 |
ENST00000555433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007027.hg.1 |
ENST00000555433.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007027.hg.1 |
ENST00000556228.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007027.hg.1 |
ENST00000556228.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007027.hg.1 |
ENST00000557465.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007027.hg.1 |
ENST00000557465.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007027.hg.1 |
ENST00000557465.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1400007027.hg.1 |
ENST00000557465.1 |
Ensembl |