Assay Details
| Gene Symbol: C1orf61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135266.1 | 1 - 2 | 210 | 81 | ||
| GenBank mRNA | HY137784.1 | - | 1 - 2 | 210 | 81 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 9-1 | MIR9-1 |
MIRN9-1, hsa-mir-9-1, miRNA9-1, mir-9-1 |
Sense | Chr. 1 - 156420341 - 156420429 on NCBI Build GRCh38 | |
| Downstream | myocyte enhancer factor 2D | MEF2D | 42.0kb | Sense | Chr. 1 - 156463721 - 156500842 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

C1orf61
Genomic Map