Assay Details
| Gene Symbol: LOC105378183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135269.1 | 1 - 2 | 55 | 118 | ||
| GenBank mRNA | AA973324.1 | - | 1 - 2 | 117 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 605 | LINC00605 | -128.3kb | Anti-sense | Chr. 14 - 103187221 - 103189028 on NCBI Build GRCh38 | |
| Downstream | translation initiation factor IF-2-like | LOC105370687 | 13.1kb | Anti-sense | Chr. 14 - 103331672 - 103335046 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105378183
Genomic Map