Assay Details
| Gene Symbol: LOC105370619 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135274.1 | 1 - 2 | 226 | 109 | ||
| GenBank mRNA | BU568198.1 | - | 1 - 2 | 205 | 109 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 642 | LINC00642 | -56.9kb | Anti-sense | Chr. 14 - 90455230 - 90458905 on NCBI Build GRCh38 | |
| Downstream | tetratricopeptide repeat domain 7B | TTC7B |
TTC7L1, c14_5685 |
23.4kb | Sense | Chr. 14 - 90540588 - 90816479 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105370619
Genomic Map