Assay Details
| Gene Symbol: LINC00517 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135286.1 | 1 - 2 | 104 | 109 | ||
| GenBank mRNA | AK125955.1 | - | 1 - 2 | 105 | 109 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | tetratricopeptide repeat domain 6 | TTC6 |
C14orf25, NCRNA00291 |
-53.4kb | Anti-sense | Chr. 14 - 37595199 - 37842628 on NCBI Build GRCh38 |
| Downstream | somatostatin receptor 1 | SSTR1 |
SRIF-2, SS-1-R, SS1-R, SS1R |
305.9kb | Anti-sense | Chr. 14 - 38207999 - 38213063 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC00517
Genomic Map