Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007440.hg.1 |
ENST00000438190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007440.hg.1 |
ENST00000438190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007440.hg.1 |
ENST00000438190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007440.hg.1 |
ENST00000438190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007440.hg.1 |
ENST00000438190.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007440.hg.1 |
ENST00000445544.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007440.hg.1 |
ENST00000445544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007440.hg.1 |
ENST00000438190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007440.hg.1 |
ENST00000438190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007440.hg.1 |
ENST00000438190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007440.hg.1 |
ENST00000438190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007440.hg.1 |
ENST00000438190.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007440.hg.1 |
ENST00000445544.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007440.hg.1 |
ENST00000445544.1 |
Ensembl |