Assay Details
| Gene Symbol: LINC00863 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135292.1 | 2 - 3 | 463 | 88 | ||
| GenBank mRNA | BC082979.1 | - | 2 - 3 | 277 | 88 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NUTM2A antisense RNA 1 | NUTM2A-AS1 | FAM22A-AS1 | -1.9kb | Anti-sense | Chr. 10 - 87238667 - 87342558 on NCBI Build GRCh38 |
| Downstream | NUT family member 2D | NUTM2D | FAM22D | 2.8kb | Sense | Chr. 10 - 87357720 - 87370695 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC00863
Genomic Map