Assay Details
| Gene Symbol: LOC105374177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135546.1 | 1 - 2 | 1083 | 101 | ||
| GenBank mRNA | BC038760.1 | - | 1 - 2 | 1081 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
leucine, glutamate and lysine rich 1 |
LEKR1 | -34.2kb | Sense | Chr. 3 - 156825228 - 157047659 on NCBI Build GRCh38 | |
| Downstream | long intergenic non-protein coding RNA 881 | LINC00881 | 1.9kb | Sense | Chr. 3 - 157089881 - 157101135 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105374177
Genomic Map