Assay Details
| Gene Symbol: LOC101929882 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135558.1 | 1 - 2 | 98 | 93 | ||
| GenBank mRNA | DA578706.1 | - | 1 - 2 | 98 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | grainyhead like transcription factor 1 | GRHL1 |
LBP32, MGR, NH32, TFCP2L2 |
-40.3kb | Anti-sense | Chr. 2 - 9951663 - 10002284 on NCBI Build GRCh38 |
| Downstream | Kruppel like factor 11 | KLF11 |
FKLF, FKLF1, MODY7, TIEG2, Tieg3 |
154.0 | Anti-sense | Chr. 2 - 10043555 - 10054836 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101929882
Genomic Map