Assay Details
| Gene Symbol: LOC100996630 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135588.1 | 1 - 2 | 117 | 96 | ||
| GenBank mRNA | AI018412.1 | - | 1 - 2 | 96 | ||
| BX417179.2 | - | 1 - 2 | 117 | 96 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RWD domain containing 3 | RWDD3 | RSUME | -63.7kb | Anti-sense | Chr. 1 - 95234155 - 95247225 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC440594 | FLJ31662 | 156.5kb | Anti-sense | Chr. 1 - 95474737 - 95479356 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC100996630
Genomic Map