Assay Details
| Gene Symbol: LOC101927727 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135633.1 | 1 - 2 | 302 | 69 | ||
| GenBank mRNA | BG617385.1 | - | 1 - 2 | 58 | 69 | |
| BQ934941.1 | - | 1 - 2 | 302 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nucleoredoxin | NXN |
NRX, TRG-4 |
Anti-sense | Chr. 17 - 799313 - 979766 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC100130876 | LOC100130876 | 29.1kb | Anti-sense | Chr. 17 - 940340 - 943610 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101927727
Genomic Map