Assay Details
| Gene Symbol: LOC105371485 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135642.1 | 1 - 2 | 391 | 111 | ||
| GenBank mRNA | BC040634.1 | - | 1 - 2 | 224 | 111 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | reticulon 4 receptor like 1 | RTN4RL1 |
NGRH2, NgR3 |
Anti-sense | Chr. 17 - 1934677 - 2025345 on NCBI Build GRCh38 | |
| Downstream | diphthamide biosynthesis 1 | DPH1 |
DEDSSH, DPH2L, DPH2L1, OVCA1 |
9.4kb | Sense | Chr. 17 - 2030137 - 2043431 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105371485
Genomic Map