Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000525111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000525111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000525111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000525111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000525111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000529264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000529264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000529264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000529264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000529264.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000532387.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000532387.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000532387.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000532387.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000532387.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000612281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000612281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000612281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000612281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000612281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000614277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000614277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000614277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000614277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000614277.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000621428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000621428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000621428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000621428.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1700012394.hg.1 |
ENST00000621428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000525111.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000525111.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000525111.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000525111.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000525111.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000529264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000529264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000529264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000529264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000529264.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000532387.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000532387.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000532387.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000532387.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000532387.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000612281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000612281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000612281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000612281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000612281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000614277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000614277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000614277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000614277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000614277.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000621428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000621428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000621428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000621428.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1700012394.hg.1 |
ENST00000621428.1 |
Ensembl |