Assay Details
| Gene Symbol: LOC105371824 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135675.1 | 2 - 3 | 396 | 88 | ||
| GenBank mRNA | BU678252.1 | - | 1 - 2 | 88 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | MYCBP associated protein | MYCBPAP |
AMAP-1, AMAP1 |
-5.8kb | Anti-sense | Chr. 17 - 50507778 - 50531502 on NCBI Build GRCh38 |
| Downstream | spermatogenesis associated 20 | SPATA20 |
HEL-S-98, SSP411, Tisp78 |
8.9kb | Anti-sense | Chr. 17 - 50547089 - 50555852 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105371824
Genomic Map