Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008942.hg.1 |
ENST00000576873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008942.hg.1 |
ENST00000576873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008942.hg.1 |
ENST00000576873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008942.hg.1 |
ENST00000576873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008942.hg.1 |
ENST00000576873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008942.hg.1 |
ENST00000576873.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008942.hg.1 |
ENST00000613733.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008942.hg.1 |
ENST00000613733.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008942.hg.1 |
ENST00000613733.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008942.hg.1 |
ENST00000613931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500008942.hg.1 |
ENST00000613931.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009968.hg.1 |
ENST00000565305.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009968.hg.1 |
ENST00000565305.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009968.hg.1 |
ENST00000565305.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009968.hg.1 |
ENST00000565305.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009968.hg.1 |
ENST00000565305.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009968.hg.1 |
ENST00000565305.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009968.hg.1 |
ENST00000565305.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009968.hg.1 |
ENST00000565305.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009968.hg.1 |
ENST00000565543.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009968.hg.1 |
ENST00000565543.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009968.hg.1 |
ENST00000565543.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009968.hg.1 |
ENST00000565543.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500009968.hg.1 |
ENST00000565543.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008942.hg.1 |
ENST00000576873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008942.hg.1 |
ENST00000576873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008942.hg.1 |
ENST00000576873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008942.hg.1 |
ENST00000576873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008942.hg.1 |
ENST00000576873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008942.hg.1 |
ENST00000576873.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008942.hg.1 |
ENST00000613733.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008942.hg.1 |
ENST00000613733.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008942.hg.1 |
ENST00000613733.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008942.hg.1 |
ENST00000613931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500008942.hg.1 |
ENST00000613931.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009968.hg.1 |
ENST00000565305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009968.hg.1 |
ENST00000565305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009968.hg.1 |
ENST00000565305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009968.hg.1 |
ENST00000565305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009968.hg.1 |
ENST00000565305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009968.hg.1 |
ENST00000565305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009968.hg.1 |
ENST00000565305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009968.hg.1 |
ENST00000565305.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009968.hg.1 |
ENST00000565543.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009968.hg.1 |
ENST00000565543.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009968.hg.1 |
ENST00000565543.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009968.hg.1 |
ENST00000565543.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500009968.hg.1 |
ENST00000565543.1 |
Ensembl |