Assay Details
| Gene Symbol: LOC101929151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135691.1 | 1 - 2 | 726 | 59 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transducin like enhancer of split 3 | TLE3 |
ESG, ESG3, GRG3, HsT18976 |
-473.0kb | Sense | Chr. 15 - 70047790 - 70097935 on NCBI Build GRCh38 |
| Downstream | senescence associated long non-coding RNA 3 | SALRNA3 |
SAL-RNA3, XLOC_025918 |
28.9kb | Anti-sense | Chr. 15 - 70615547 - 70616567 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101929151
Genomic Map