Assay Details
| Gene Symbol: LOC105370369 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135814.1 | 1 - 2 | 41 | 72 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC107983958 | LOC107983958 | Sense | Chr. 13 - 111623249 - 111637452 on NCBI Build GRCh38 | ||
| Downstream | long intergenic non-protein coding RNA 354 | LINC00354 | 251.3kb | Sense | Chr. 13 - 111893378 - 111901176 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105370369
Genomic Map