Assay Details
| Gene Symbol: LOC105378828 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_135837.1 | 2 - 3 | 947 | 117 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
chloride channel accessory 3, pseudogene |
CLCA3P | CLCA3 | -41.6kb | Anti-sense | Chr. 1 - 86634276 - 86655376 on NCBI Build GRCh38 |
| Downstream | SH3 domain containing GRB2 like endophilin B1 | SH3GLB1 |
Bif-1, CGI-61, PPP1R70, dJ612B15.2 |
6.8kb | Anti-sense | Chr. 1 - 86704570 - 86748184 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105378828
Genomic Map