Assay Details
| Gene Symbol: LINC00841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033846.2 | 1 - 2 | 73 | 76 | ||
| NR_136147.1 | 1 - 2 | 73 | 76 | |||
| GenBank mRNA | AK310302.1 | - | 1 - 2 | 65 | 76 | |
| HY016364.1 | - | 1 - 2 | 73 | 76 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 840 | LINC00840 | -13.9kb | Sense | Chr. 10 - 43859411 - 43895433 on NCBI Build GRCh38 | |
| Downstream | C-X-C motif chemokine ligand 12 | CXCL12 |
IRH, PBSF, SCYB12, SDF1, TLSF, TPAR1 |
382.4kb | Anti-sense | Chr. 10 - 44292088 - 44385097 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC00841
Genomic Map