Assay Details
| Gene Symbol: LOC101060391 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136151.1 | 2 - 3 | 589 | 109 | ||
| GenBank mRNA | CD519625.1 | - | 1 - 2 | 387 | 109 | |
| DB544287.1 | - | 3 - 4 | 331 | 109 | ||
| DB545731.1 | - | 3 - 4 | 343 | 109 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101060385 | LOC101060385 | -44.2kb | Sense | Chr. 2 - 900216 - 905451 on NCBI Build GRCh38 | |
| Downstream | syntrophin gamma 2 | SNTG2 |
G2SYN, SYN5 |
482.0 | Anti-sense | Chr. 2 - 950868 - 1367991 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101060391
Genomic Map