Assay Details
| Gene Symbol: LOC100128905 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136175.1 | 1 | 1625 | 68 | ||
| GenBank mRNA | AI677644.1 | - | 1 | 68 | ||
| BC092496.1 | - | 1 | 1489 | 68 | ||
| BX091417.1 | - | 1 | 68 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Sp3 transcription factor | SP3 | SPR2 | -59.6kb | Anti-sense | Chr. 2 - 173906459 - 173965702 on NCBI Build GRCh38 |
| Downstream | Obg-like ATPase 1 | OLA1 |
DOC45, GBP45, GTBP9, GTPBP9, PTD004 |
45.3kb | Anti-sense | Chr. 2 - 174072447 - 174248637 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC100128905
Genomic Map