Assay Details
| Gene Symbol: LOC101929163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136244.1 | 2 - 3 | 435 | 71 | ||
| GenBank mRNA | BM709398.1 | - | 2 - 3 | 435 | 71 | |
| DA342640.1 | - | 2 - 3 | 253 | 71 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 6 open reading frame 10 | C6orf10 | TSBP | Anti-sense | Chr. 6 - 32290523 - 32371912 on NCBI Build GRCh38 | |
| Downstream | HLA complex group 23 (non-protein coding) | HCG23 | dJ1077I5.3 | 24.7kb | Sense | Chr. 6 - 32390510 - 32393691 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101929163
Genomic Map