Assay Details
| Gene Symbol: LOC105377879 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136251.1 | 1 - 2 | 280 | 106 | ||
| GenBank mRNA | HY045387.1 | - | 1 - 2 | 231 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | synaptosome associated protein 91 | SNAP91 |
AP180, CALM |
-34.0 | Anti-sense | Chr. 6 - 83552885 - 83709408 on NCBI Build GRCh38 |
| Downstream | ripply transcriptional repressor 2 | RIPPLY2 |
C6orf159, SCDO6, dJ237I15.1 |
130.1kb | Sense | Chr. 6 - 83853266 - 83857515 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105377879
Genomic Map