Assay Details
| Gene Symbol: LOC105377879 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136251.1 | 2 - 3 | 357 | 105 | ||
| GenBank mRNA | HY045387.1 | - | 2 - 3 | 308 | 105 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | synaptosome associated protein 91 | SNAP91 |
AP180, CALM |
-13.6kb | Anti-sense | Chr. 6 - 83552885 - 83709408 on NCBI Build GRCh38 |
| Downstream | ripply transcriptional repressor 2 | RIPPLY2 |
C6orf159, SCDO6, dJ237I15.1 |
126.8kb | Sense | Chr. 6 - 83853266 - 83857515 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105377879
Genomic Map