Assay Details
| Gene Symbol: LOC101927420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136253.1 | 1 - 2 | 191 | 78 | ||
| GenBank mRNA | AA527254.1 | - | 1 - 2 | 117 | 78 | |
| HY287918.1 | - | 1 - 2 | 78 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | KIAA1324 like | KIAA1324L | EIG121L | -91.7kb | Sense | Chr. 7 - 86876906 - 87059714 on NCBI Build GRCh38 |
| Downstream | transmembrane protein 243 | TMEM243 |
C7orf23, MM-TRAG, MMTRAG |
43.1kb | Sense | Chr. 7 - 87195593 - 87220603 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC101927420
Genomic Map