Assay Details
| Gene Symbol: LOC105374820 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_136311.1 | 2 - 3 | 144 | 109 | ||
| GenBank mRNA | AA758420.1 | - | 1 - 2 | 109 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101927967 | LOC101927967 | -261.9kb | Anti-sense | Chr. 2 - 77743696 - 78290731 on NCBI Build GRCh38 | |
| Downstream | regenerating family member 3 gamma | REG3G |
LPPM429, PAP IB, PAP-1B, PAP1B, PAPIB, REG III, REG-III, UNQ429 |
459.6kb | Sense | Chr. 2 - 79025686 - 79028504 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LOC105374820
Genomic Map